Learn More
Novus Biologicals™ Mouse Bcl-2 ELISA Kit (Colorimetric)
SDP
ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture.
Supplier:  Novus Biologicals™ NBP269946
Catalog No. NBP269946
Add to Cart
Description
Description
The Mouse Bcl-2 ELISA Kit (Colorimetric) measures mouse Bcl-2 in serum, plasma and other biological fluids.
Specifications
Specifications
15.63 - 1000 pg/mL
This kit recognizes Mouse Bcl-2 in samples. No significant cross-reactivity or interference between Mouse Bcl-2 and analogues was observed.
Biological Fluids, Plasma, Serum
596
Mouse
Storage of components varies. See protocol for specific instructions.
9.38 pg/mL
ELISA Kit (Colorimetric)
ELISA
BCL2
1 Kit
Bcl-2
Recommendations
Recommendations