Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Gibco™ Mouse beta-Defensin 1 Recombinant Protein, PeproTech®
Recombinant Protein
Supplier: Gibco™ 250441MG
Description
Defensins form a family of microbicidal and cytotoxic peptides made by neutrophils. Members of the defensin family are highly similar in protein sequence. This gene encodes defensin, beta 1, an antimicrobial peptide implicated in the resistance of epithelial surfaces to microbial colonization. This gene maps in close proximity to defensin family member, defensin, alpha 1 and has been implicated in the pathogenesis of cystic fibrosis.Specifications
P56386 | |
Lyophilized | |
13214 | |
DEFB1 | |
1mg | |
RUO | |
AW260221; BD1; BD-1; beta defensin 1; beta-defensin 1; beta-defensin-1; DEFB1; DEFB-1; DEFB101; defensin beta 1; defensin, beta 1; HBD1; rBD-1 | |
Protein | |
Unconjugated | |
DQYKCLQHGG FCLRSSCPSN TKLQGTCKPD KPNCCKS | |
≥98% by SDS-PAGE gel and HPLC analyses. |
Functional Assay | |
Protein with no preservative | |
4.1 kDa | |
HPLC, SDS-PAGE | |
-20°C | |
<1 EU/μg | |
DEFB1 | |
Mouse | |
Recombinant | |
E. coli |
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction