Learn More
Novus Biologicals™ Mouse BMP-7 ELISA Kit (Colorimetric)
ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture.
Manufacturer:  Novus Biologicals™ NBP270000
Catalog No. NBP270000
Add to cart
Description
Description
The Mouse BMP-7 ELISA Kit (Colorimetric) measures mouse BMP-7 in serum, plasma and other biological fluids.
Specifications
Specifications
15.63 - 1000 pg/mL
ELISA
ELISA Kit (Colorimetric)
Storage is content dependent.
Mouse
1 Kit
This kit recognizes Mouse BMP-7 in samples. No significant cross-reactivity or interference between Mouse BMP-7 and analogues was observed.
655
9.38 pg/mL
BMP-7
BMP7
Biological Fluids, Plasma, Serum