Learn More
Novus Biologicals Mouse BMP-8b ELISA Kit (Colorimetric)
ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture.
Manufacturer: Novus Biologicals NBP270003
Description
The Mouse BMP-8b ELISA Kit (Colorimetric) measures mouse BMP-8b in serum, plasma and other biological fluids.Specifications
15.63 - 1000 pg/mL | |
ELISA | |
ELISA Kit (Colorimetric) | |
Storage is content dependent. | |
Mouse | |
1 Kit |
This kit recognizes Mouse BMP8B in samples. No significant cross-reactivity or interference between Mouse BMP8B and analogues was observed. | |
656 | |
9.38 pg/mL | |
BMP-8b | |
BMP8B | |
Biological Fluids, Plasma, Serum |