Learn More
Novus Biologicals™ Mouse CACYBP ELISA Kit (Colorimetric)
ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture.
Manufacturer:  Novus Biologicals™ NBP274963
Catalog No. NBP274963
Add to cart
Description
Description
The Mouse CACYBP ELISA Kit (Colorimetric) measures mouse CACYBP in serum, plasma and other biological fluids.
Specifications
Specifications
15.63 - 1000 pg/mL
ELISA
ELISA Kit (Colorimetric)
Storage is content dependent.
Mouse
1 Kit
This kit recognizes Mouse CACYBP in samples. No significant cross-reactivity or interference between Mouse CACYBP and analogues was observed.
27101
9.38 pg/mL
CACYBP
CACYBP
Biological Fluids, Plasma, Serum