Learn More
Abnova™ Mouse Calb2 (Q08331) Full-length Recombinant Protein
Used for SDS-PAGE
Manufacturer:  Abnova™ P4483
Catalog No. 89-937-828
Add to cart
Description
Description
Specifications
Specifications
Q08331
Liquid
12308
Calb2 (Mouse) Recombinant Protein
5 ug
RUO
CR
Calb2
Yes
Sodium Dodecyl Sulfate–Polyacrylamide Gel Electrophoresis
Liquid
57kDa
≥90% by SDS-PAGE and HPLC
Store at -20°C. Aliquot to avoid repeated freezing and thawing.
<0.1ng/μg
Calb2
Escherichia coli
None