Learn More
BioworldSupplier Diversity Partner MOUSE CALRETININ (RECOMBINANT) DFS Item
Manufacturer:  BioworldSupplier Diversity Partner 220605841
Recombinant Murine Calretinin Source: E.Coli Background: Calretinin is an intracellular calcium-binding protein belonging to the troponin C superfamily characterized by a structural mitif described as the EF-hand domain. The immunohi...
Catalog No. 50-253-684