Learn More
Novus Biologicals™ Mouse Calumenin ELISA Kit (Colorimetric)
ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture.
Manufacturer:  Novus Biologicals™ NBP274972
Catalog No. NBP274972
Add to cart
Description
Description
The Mouse Calumenin ELISA Kit (Colorimetric) measures mouse Calumenin in serum, plasma and other biological fluids.
Specifications
Specifications
7.81 - 500 pg/mL
ELISA
ELISA Kit (Colorimetric)
Storage is content dependent.
Mouse
1 Kit
This kit recognizes Mouse CALU in samples. No significant cross-reactivity or interference between Mouse CALU and analogues was observed.
813
4.69 pg/mL
Calumenin
CALU
Biological Fluids, Plasma, Serum