Learn More
Novus Biologicals Mouse Cathepsin B ELISA Kit (Colorimetric)
ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture.
Manufacturer: Novus Biologicals NBP267254
Description
The Mouse Cathepsin B ELISA Kit (Colorimetric) measures mouse Cathepsin B in serum, plasma and other biological fluids.Specifications
ELISA Kit (Colorimetric) | |
This kit recognizes Mouse CTSB in samples. No significant cross-reactivity or interference between Mouse CTSB and analogues was observed. | |
1508 | |
Cathepsin B | |
1 Kit | |
0.10 ng/mL |
0.16 - 10 ng/mL | |
ELISA | |
Storage is content dependent. | |
Mouse | |
Biological Fluids, Plasma, Serum | |
CTSB |