Learn More
Raybiotech IncSupplier Diversity Partner Mouse Cathepsin E ELISA Kit DFS Item
Manufacturer:  Raybiotech IncSupplier Diversity Partner ELMCTSE1
For Serum, Plasma, and Cell Culture Supernatants. Sensitivity: 14 pg/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: This ELISA antibody pair recognizes Mouse Cathepsin E.
Catalog No. 50-194-9958