Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.
Novus Biologicals™ Mouse CCL2/MCP1 ELISA Kit [Biotin]
ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture. 
Manufacturer:  Novus Biologicals™ NBP192659
Catalog No. NBP192659
Description
Description
The Mouse CCL2/MCP1 ELISA Kit (Colorimetric) measures mouse CCL2/MCP1 in cell culture supernatant, serum.
Specifications
Specifications
15.6 - 1,000pg/mL
6347
ELISA Kit
2.2pg/mL
Mouse (Mu)
CCL2/MCP1
CCL2
ELISA
Aluminium pouch(es) with a Microwell Plate coated with monoclonal antibody to mouse MCP-1, Biotin-Conjugate anti-mouse MCP-1 monoclonal antibody, Streptavidin-HRP, Mouse MCP-1 Standard lyophilized, 2ng/mL upon reconstitution, Sample Diluent, Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA), Wash Buffer Concentrate 20x (PBS with 1% Tween 20), Substrate Solution (tetramethyl-benzidine), Stop Solution (1M Phosphoric acid), Adhesive Films
96 Tests
Cell culture supernatant, serum
Storage of components varies. See protocol for specific instructions.
Apoptosis, Cancer, Chemokine Research, Chemokines and Cytokines, Cytokine Research, Hypoxia, Immunology, Innate Immunity, Signal Transduction
C-C motif chemokine 2, chemokine (C-C motif) ligand 2, GDCF-2, HC11Monocyte secretory protein JE, MCAFMonocyte chemotactic and activating factor, MCP1 Monocyte chemotactic protein 1, MCP-1 Small-inducible cytokine A2, MGC9434, Monocyte chemoattractant pro