Learn More
Novus Biologicals™ Mouse CD4 ELISA Kit (Colorimetric)
ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture.
Supplier: Novus Biologicals™ NBP275148
Description
The Mouse CD4 ELISA Kit (Colorimetric) measures mouse CD4 in serum, plasma and other biological fluids.Specifications
0.39 - 25 ng/mL | |
This kit recognizes Mouse CD4 in samples. No significant cross-reactivity or interference between Mouse CD4 and analogues was observed. | |
Biological Fluids, Plasma, Serum | |
920 | |
Mouse | |
Adaptive Immunity, Cell Biology, Cellular Markers, Cytokine Research, Immunology, Innate Immunity, Regulatory Immunology, Signal Transduction, Stem Cell Markers | |
CD4 |
0.23 ng/mL | |
ELISA Kit (Colorimetric) | |
ELISA | |
CD4 | |
1 Kit | |
Storage of components varies. See protocol for specific instructions. |