Learn More
Novus Biologicals™ Mouse CD40 Ligand/TNFSF5 ELISA Kit (Colorimetric)
ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture.
Supplier: Novus Biologicals™ NBP275151
Description
The Mouse CD40 Ligand/TNFSF5 ELISA Kit (Colorimetric) measures mouse CD40 Ligand/TNFSF5 in serum, plasma and other biological fluids.Specifications
93.75 - 6000 pg/mL | |
This kit recognizes Mouse CD40L in samples. No significant cross-reactivity or interference between Mouse CD40L and analogues was observed. | |
Biological Fluids, Plasma, Serum | |
959 | |
Mouse | |
Storage of components varies. See protocol for specific instructions. |
56.25 pg/mL | |
ELISA Kit (Colorimetric) | |
ELISA | |
CD40LG | |
1 Kit | |
CD40 Ligand/TNFSF5 |