Learn More
Novus Biologicals™ Mouse CD40 Ligand/TNFSF5 ELISA Kit
ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture. 
Manufacturer:  Novus Biologicals™ NBP192662
Catalog No. NBP192662
Add to cart
Description
Description
The Mouse CD40 Ligand/TNFSF5 ELISA Kit (Colorimetric) measures mouse CD40 Ligand/TNFSF5 in cell culture supernatant, serum.
Specifications
Specifications
0.31 - 20.0ng/mL
959
ELISA Kit
0.14ng/mL
Mouse (Mu)
CD40 Ligand/TNFSF5
CD40LG
ELISA
Aluminium pouch(es) with a Microwell Plate coated with monoclonal antibody to mouse sCD40L, Biotin-Conjugate anti-mouse sCD40L monoclonal antibody, Streptavidin-HRP, Mouse sCD40L Standard lyophilized, 40ng/mL upon reconstitution, Sample Diluent, Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA), Wash Buffer Concentrate 20x (PBS with 1% Tween 20), Substrate Solution (tetramethyl-benzidine), Stop Solution (1M Phosphoric acid), Adhesive Films
96 Tests
Cell culture supernatant, serum
Storage of components varies. See protocol for specific instructions.
Adaptive Immunity, Apoptosis, Asthma, Breast Cancer, Cancer, Cell Biology, Cellular Markers, Endothelial Cell Markers, Hematopoietic Stem Cell Markers, Hypoxia, Immunology, Innate Immunity, Mast Cell Markers, Mesenchymal Stem Cell Markers, Myeloid Cell Ma
CD154, CD154 antigen, CD40 antigen ligand, CD40 ligand, CD40-L, CD40LIGM, gp39, hCD40L, HIGM1, T-B cell-activating molecule, T-BAM, T-cell antigen Gp39, TNF-related activation protein, TNFSF5IMD3, TRAPtumor necrosis factor (ligand) superfamily, member 5 (