Learn More
ACROBiosystems ACROBiosystems Mouse CD79B Protein, Fc Tag
Supplier:  ACROBiosystems CDBM5253100UG
Encompass_Preferred
Recombinant Protein;100UG;Mouse CD79B, Fc Tag (CDB-M5253) is expressed from human 293 cells (HEK293). It contains AA Val 26 - Asp 158 (Accession # P15530-1).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 40.3 kDa.
Catalog No. 50-210-7240