Learn More
Novus Biologicals™ Mouse CEACAM5/CD66e ELISA Kit (Colorimetric)
ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture.
Manufacturer:  Novus Biologicals™ NBP275216
Catalog No. NBP275216
Add to cart
Description
Description
The Mouse CEACAM5/CD66e ELISA Kit (Colorimetric) measures mouse CEACAM5/CD66e in serum, plasma and other biological fluids.
Specifications
Specifications
62.50 - 4000 pg/mL
ELISA
ELISA Kit (Colorimetric)
Storage is content dependent.
Mouse
1 Kit
This kit recognizes Mouse CEA in samples. No significant cross-reactivity or interference between Mouse CEA and analogues was observed.
1048
37.50 pg/mL
CEACAM5/CD66e
CEACAM5
Biological Fluids, Plasma, Serum