Learn More
Novus Biologicals™ Mouse Cklfsf8 ELISA Kit (Colorimetric)
ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture.
Supplier: Novus Biologicals™ NBP275304
Description
The Mouse Cklfsf8 ELISA Kit (Colorimetric) measures mouse Cklfsf8 in serum, plasma and other biological fluids.Specifications
0.16 - 10 ng/mL | |
This kit recognizes Mouse CKLFSF8 in samples. No significant cross-reactivity or interference between Mouse CKLFSF8 and analogues was observed. | |
Biological Fluids, Plasma, Serum | |
152189 | |
Mouse | |
Storage of components varies. See protocol for specific instructions. |
0.10 ng/mL | |
ELISA Kit (Colorimetric) | |
ELISA | |
CMTM8 | |
1 Kit | |
Cklfsf8 |