Learn More
Crystal Chem IncSupplier Diversity Partner Mouse Clusterin ELISA Kit, 96-wells DFS Item
Manufacturer:  Crystal Chem IncSupplier Diversity Partner 80633
The Mouse Clusterin ELISA is used to quantify levels of mouse clusterin in serum and plasma using only a 2 µL sample volume. Highlights: Kit uses sample size of only 2 µL. High sensitivity of 1.1 ng/mL. Dynamic range of 1.56 - 100 ng/mL.Crystal Chem is a worldwide leading supplier of mouse and rat ELISA kits for research use. Our kits are among the highest quality found in the market, and are used by leading research universities, pharma companies, and government institutions around the world.
Catalog No. 50-194-7764