Learn More
Novus Biologicals™ Mouse Collagen IV ELISA Kit (Colorimetric)
ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture.
Manufacturer:  Novus Biologicals™ NBP275865
Catalog No. NBP275865
Add to cart
Description
Description
The Mouse Collagen IV ELISA Kit (Colorimetric) measures mouse Collagen IV in serum, plasma and other biological fluids.
Specifications
Specifications
1.56 -100 ng/mL
ELISA
0.94 ng/mL
Collagen IV
1 Kit
This kit recognizes Mouse COL4 in samples. No significant cross-reactivity or interference between Mouse COL4 and analogues was observed.
ELISA Kit (Colorimetric)
Storage is content dependent.
Mouse
Biological Fluids, Plasma, Serum