Learn More
Novus Biologicals™ Mouse Collagen X ELISA Kit (Colorimetric)
ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture.
Manufacturer:  Novus Biologicals™ NBP275827
Catalog No. NBP275827
Add to cart
Description
Description
The Mouse Collagen X ELISA Kit (Colorimetric) measures mouse Collagen X in serum, plasma and other biological fluids.
Specifications
Specifications
0.31 - 20 ng/mL
ELISA
0.19 ng/mL
Collagen X
1 Kit
This kit recognizes Mouse COL10 in samples. No significant cross-reactivity or interference between Mouse COL10 and analogues was observed.
ELISA Kit (Colorimetric)
Storage is content dependent.
Mouse
Biological Fluids, Plasma, Serum