Learn More
Novus Biologicals™ Mouse Complement Factor B ELISA Kit (Colorimetric)
ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture.
Supplier: Novus Biologicals™ NBP275243
Description
The Mouse Complement Factor B ELISA Kit (Colorimetric) measures mouse Complement Factor B in serum, plasma and other biological fluids.Specifications
1.56 - 100 ng/mL | |
This kit recognizes Mouse CFB in samples. No significant cross-reactivity or interference between Mouse CFB and analogues was observed. | |
Biological Fluids, Plasma, Serum | |
629 | |
Mouse | |
Immunology | |
BF AHUS4, B-factor, properdin, BFD, C3 proaccelerator, C3 proactivator, C3/C5 convertase, complement factor B, EC 3.4.21, EC 3.4.21.47, FB, FBI12, GBGCFAB, Glycine-rich beta glycoprotein, glycine-rich beta-glycoprotein, H2-Bf, PBF2, Properdin factor B |
0.94 ng/mL | |
ELISA Kit (Colorimetric) | |
ELISA | |
CFB | |
1 Kit | |
Storage of components varies. See protocol for specific instructions. | |
Complement Factor B |