Learn More
Novus Biologicals™ Mouse Complement Factor H ELISA Kit (Colorimetric)
ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture.
Manufacturer:  Novus Biologicals™ NBP267239
Catalog No. NBP267239
Add to cart
Description
Description
The Mouse Complement Factor H ELISA Kit (Colorimetric) measures mouse Complement Factor H in serum, plasma and other biological fluids.
Specifications
Specifications
15.63 - 1000 ng/mL
This kit recognizes Mouse CFH in samples. No significant cross-reactivity or interference between Mouse CFH and analogues was observed.
3075
Mouse
1 Kit
Storage is content dependent.
9.38 ng/mL
ELISA
CFH
ELISA Kit (Colorimetric)
Biological Fluids,Plasma,Serum
Complement Factor H