Learn More
Novus Biologicals™ Mouse Contactin-3 ELISA Kit (Colorimetric)
ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture.
Manufacturer:  Novus Biologicals™ NBP275808
Catalog No. NBP275808
Add to cart
Description
Description
The Mouse Contactin-3 ELISA Kit (Colorimetric) measures mouse Contactin-3 in serum, plasma and other biological fluids.
Specifications
Specifications
0.16 - 10 ng/mL
ELISA
ELISA Kit (Colorimetric)
Storage is content dependent.
Mouse
1 Kit
This kit recognizes Mouse CNTN3 in samples. No significant cross-reactivity or interference between Mouse CNTN3 and analogues was observed.
5067
0.10 ng/mL
Contactin-3
CNTN3
Biological Fluids, Plasma, Serum