Learn More
Abnova™ Mouse Csf3 (P09920) Recombinant Protein
Used for Func, SDS-PAGE
Manufacturer:  Abnova™ P4594
Catalog No. 89-940-713
Add to cart
Description
Description
Sequence: MVPLVTVSALPPSLPLPRSFLLKSLEQVRKIQASGSVLLEQLCATYKLCHPEELVLLGHSLGIPKASLSGCSSQALQQTQCLSQLHSGLCLYQGLLQALSGISPALAPTLDLLQLDVANFATTIWQQMENLGVAPTVQPTQSAMPAFTSAFQRRAGGVLAISYLQGFLETARLALHHLA
Specifications
Specifications
P09920
Lyophilized
12985
Csf3 (Mouse) Recombinant Protein
1 ug/lane in 4-20% Tris-Glycine gel Stained with Coomassie Blue
MVPLVTVSALPPSLPLPRSFLLKSLEQVRKIQASGSVLLEQLCATYKLCHPEELVLLGHSLGIPKASLSGCSSQALQQTQCLSQLHSGLCLYQGLLQALSGISPALAPTLDLLQLDVANFATTIWQQMENLGVAPTVQPTQSAMPAFTSAFQRRAGGVLAISYLQGFLETARLALHHLA
RUO
Csfg|G-CSF|MGI-IG
Csf3
The activity is determined by the dose-dependent proliferation of mouse M-NFS-60 cells. The expected ED50 for this effect is 8-12pg/mL.
Yes
Functional Study, Sodium Dodecyl Sulfate–Polyacrylamide Gel Electrophoresis
Lyophilized
19kDa
Escherichia coli expression system
10 ug
Store at -20°C on dry atmosphere. After reconstitution with sterilized water, store at -20°C. Aliquot to avoid repeated freezing and thawing.
<0.1 EU/μg
Csf3
Escherichia coli expression system
Escherichia coli
None