Learn More
Abnova™ Mouse Cxcl1 (P12850) Recombinant Protein
Used for Func, SDS-PAGE
Manufacturer:  Abnova™ P4603
Catalog No. 89-941-032
Add to cart
Description
Description
Sequence: APIANELRCQCLQTMAGIHLKNIQSLKVLPSGPHCTQTEVIATLKNGREACLDPEAPLVQKIVQKMLKGVPK
Specifications
Specifications
P12850
Lyophilized
14825
Cxcl1 (Mouse) Recombinant Protein
1 ug/lane in 4-20% Tris-Glycine gel Stained with Coomassie Blue
APIANELRCQCLQTMAGIHLKNIQSLKVLPSGPHCTQTEVIATLKNGREACLDPEAPLVQKIVQKMLKGVPK
RUO
Fsp|Gro1|KC|Mgsa|N51|Scyb1|gro
Cxcl1
The activity is determined by the ability to chemoattract human neutrophils cells and is detectable starting at 10ng/mL.
Yes
Functional Study, Sodium Dodecyl Sulfate–Polyacrylamide Gel Electrophoresis
Lyophilized
7.8kDa
Escherichia coli expression system
20 ug
Store at -20°C on dry atmosphere. After reconstitution with sterilized water, store at -20°C. Aliquot to avoid repeated freezing and thawing.
<0.1 EU/μg
Cxcl1
Escherichia coli expression system
Escherichia coli
None