Learn More
Abnova™ Mouse Cxcl12 (beta) (P40224-2) Recombinant Protein
Used for Func, SDS-PAGE
Manufacturer:  Abnova™ P4628
Catalog No. 89-941-051
Add to cart
Description
Description
Sequence: KPVSLSYRCPCRFFESHIARANVKHLKILNTPNCALQIVARLKNNNRQVCIDPKLKWIQEYLEKALNKRLKM
Specifications
Specifications
P40224-2
Lyophilized
20315
Cxcl12 (Beta) (Mouse) Recombinant Protein
1 ug/lane in 4-20% Tris-Glycine gel Stained with Coomassie Blue
KPVSLSYRCPCRFFESHIARANVKHLKILNTPNCALQIVARLKNNNRQVCIDPKLKWIQEYLEKALNKRLKM
RUO
AI174028|PBSF|PBSF/SDF-1|SDF-1|Scyb12|Sdf1|Sdf1a|Sdf1b|TLSF|TLSF-a|TLSF-b|TPAR1
Cxcl12
The activity is determined by the ability to chemoattract human peripheral T cells and is detectable at 5-100ng/mL.
Yes
Functional Study, Sodium Dodecyl Sulfate–Polyacrylamide Gel Electrophoresis
Lyophilized
8.5kDa
Escherichia coli expression system
10 ug
Store at -20°C on dry atmosphere. After reconstitution with sterilized water, store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
<0.1 EU/μg
Cxcl12
Escherichia coli expression system
Escherichia coli
None