Learn More
Novus Biologicals™ Mouse CXCL14/BRAK ELISA Kit (Colorimetric)
SDP
ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture.
Supplier:  Novus Biologicals™ NBP270015
Catalog No. NBP270015
Add to Cart
Description
Description
The Mouse CXCL14/BRAK ELISA Kit (Colorimetric) measures mouse CXCL14/BRAK in serum, plasma and other biological fluids.
Specifications
Specifications
0.16 - 10 ng/mL
This kit recognizes Mouse BRAK/CXCL14 in samples. No significant cross-reactivity or interference between Mouse BRAK/CXCL14 and analogues was observed.
Biological Fluids, Plasma, Serum
9547
Mouse
Storage of components varies. See protocol for specific instructions.
0.10 ng/mL
ELISA Kit (Colorimetric)
ELISA
CXCL14
1 Kit
CXCL14/BRAK
Recommendations
Recommendations