Learn More
Mouse CXCL2 (aa 28-100), Recombinant Protein, Invitrogen™
E. coli Recombinant Protein
Manufacturer:  Thermo Scientific RP87877
Encompass
 View more versions of this product
Catalog No. PIRP87877
Add to cart
Description
Description
Produced by activated monocytes and neutrophils and expressed at sites of inflammation. Hematoregulatory chemokine, which, in vitro, suppresses hematopoietic progenitor cell proliferation. GRO-beta(5-73) shows a highly enhanced hematopoietic activity.
Specifications
Specifications
P10889
Control, ELISA, Western Blot
PBS with no preservative
7.8 kD
>95%
E. coli
RUO
C Cmotif chemokine; C x C motif chemokine; CC motif chemokine; CCmotif chemokine; chemokine (C-X-C motif) ligand 2; CINC-2 A; Cinc3; CINC-3; CXC; CXC motif chemokine; C-X-C motif chemokine 2; C-X-C motif chemokine ligand 2; CXCL; Cxcl2; cytokine-induced neutrophil chemoattractant 3; GRO b; gro beta; GRO ß; GRO2; GRO2 oncogene; GROB; GRObeta; Gro-beta; GRO-beta(5-73); GRO-beta-T; Growth-regulated protein beta; GROß; Hematopoietic synergistic factor; HSF; Macrophage inflammatory protein 2; macrophage inflammatory protein 2-alpha; melanoma growth stimulatory activity beta; MGSA beta; MGSA-b; MIP 2 A; MIP 2 alpha; MIP 2 A; Mip2; MIP-2; MIP2 a; MIP2 a; MIP2A; MIP-2 A; MIP2-alpha; SB-251353; Scyb; SCYB2; small inducible cytokine subfamily, member 2
Mouse
Yes
200 µg/mL
Liquid
20310
Mouse CXCL2 (aa 28-100)
25 µg
-20°C
0.01 ng/ µg cytokine
Cxcl2
Unlabeled