Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse CXCL2 (aa 28-100), Recombinant Protein, Invitrogen™ Non-distribution product as customer accommodation.

E. coli Recombinant Protein

$160.50 - $1959.00

Specifications

Recombinant Yes
Name Mouse CXCL2 (aa 28-100)
For Use With (Application) Control, ELISA, Western Blot
Concentration 200 µg/mL
Conjugate Unlabeled
View More Specs

Products 4
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
PIRP87876 Non-distribution product as customer accommodation.
View Documents
Thermo Scientific
RP87876
10 µg
Each for $160.50
Add to cart
 
PIRP87877 Non-distribution product as customer accommodation.
View Documents
Thermo Scientific
RP87877
25 µg
Each for $276.50
Add to cart
 
PIRP87878 Non-distribution product as customer accommodation.
View Documents
Thermo Scientific
RP87878
100 µg
Each for $751.50
Add to cart
 
PIRP87879 Non-distribution product as customer accommodation.
View Documents
Thermo Scientific
RP87879
500 µg
Each for $1,959.00
Add to cart
 
Description

Description

Produced by activated monocytes and neutrophils and expressed at sites of inflammation. Hematoregulatory chemokine, which, in vitro, suppresses hematopoietic progenitor cell proliferation. GRO-beta(5-73) shows a highly enhanced hematopoietic activity.
Specifications

Specifications

Yes
Control, ELISA, Western Blot
Unlabeled
PBS with no preservative
E. coli
-20°C
P10889
Cxcl2
>95%
Mouse CXCL2 (aa 28-100)
200 µg/mL
Liquid
C Cmotif chemokine; C x C motif chemokine; CC motif chemokine; CCmotif chemokine; chemokine (C-X-C motif) ligand 2; CINC-2 A; Cinc3; CINC-3; CXC; CXC motif chemokine; C-X-C motif chemokine 2; C-X-C motif chemokine ligand 2; CXCL; Cxcl2; cytokine-induced neutrophil chemoattractant 3; GRO b; gro beta; GRO ß; GRO2; GRO2 oncogene; GROB; GRObeta; Gro-beta; GRO-beta(5-73); GRO-beta-T; Growth-regulated protein beta; GROß; Hematopoietic synergistic factor; HSF; Macrophage inflammatory protein 2; macrophage inflammatory protein 2-alpha; melanoma growth stimulatory activity beta; MGSA beta; MGSA-b; MIP 2 A; MIP 2 alpha; MIP 2 A; Mip2; MIP-2; MIP2 a; MIP2 a; MIP2A; MIP-2 A; MIP2-alpha; SB-251353; Scyb; SCYB2; small inducible cytokine subfamily, member 2
RUO
Mouse
20310
7.8 kD
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit