Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Mouse CXCL4/PF4 ELISA Kit (Colorimetric)

Specific and sensitive quantitative evaluation of mouse PF-4 levels.

$602.00

Specifications

Species Mouse
Sensitivity 1.603ng/mL
Sample Type Serum, plasma, tissue homogenates
Research Discipline Cardiovascular Biology
Synonym chemokine (C-X-C motif) Ligand 4, C-X-C motif chemokine 4, CXCL4iroplact, Iroplact, MGC138298, Oncostatin-A, PF-4, platelet factor 4, SCYB4oncostatin-A
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB599601KT View Documents Novus Biologicals™
NBP2599601KIT
1 Kit Each for $602.00
Add to cart
 
Description

Description

The Mouse CXCL4/PF4 ELISA Kit (Colorimetric) measures mouse CXCL4/PF4 in serum, plasma, tissue homogenates.
Specifications

Specifications

Mouse
Serum, plasma, tissue homogenates
chemokine (C-X-C motif) Ligand 4, C-X-C motif chemokine 4, CXCL4iroplact, Iroplact, MGC138298, Oncostatin-A, PF-4, platelet factor 4, SCYB4oncostatin-A
CXCL4/PF4
5196
ELISA Kit (Colorimetric)
50 to 100μL
1.603ng/mL
Cardiovascular Biology
Store the unopened product at 2° to 8°C Protect from light. Do not use past expiration date.
ELISA
PF4
3.125 to 200ng/mL

For Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit