Learn More
Abnova™ Mouse Cxcl5 (P50228) Partial Recombinant Protein
Used for Func, SDS-PAGE
Manufacturer:  Abnova™ P4501
Catalog No. 89-941-079
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Specifications
Specifications
P50228
Lyophilized
20311
Mouse Cxcl5 (P50228) Partial Recombinant Protein
20μg
RUO
CXCL5
Determined by its ability to chemoattract human peripheral blood neutrophils using a concentration range of 10.0-100.0ng/mL.
Yes
Func, SDS-PAGE
Lyophilized from 20mM sodium phosphate, 150mM NaCl, pH 7.4
9.8kDa
≥97% by SDS-PAGE and HPLC
Store at −20°C on dry atmosphere.After reconstitution with sterilized ddH2O, store at −20°C.Aliquot to avoid repeated freezing and thawing.
AMCF-II, ENA-78, GCP-2, LIX, Scyb5, Scyb6
Cxcl5
E. coli
Untagged