Learn More
Novus Biologicals™ Mouse Cyclophilin-F ELISA Kit (Colorimetric)
Colorimetric ELISA Kit
Supplier: Novus Biologicals™ NBP306739
This item is not returnable.
View return policy
Specifications
0.31 to 20 ng/mL | |
ELISA Kit (Colorimetric) | |
This kit recognizes Mouse Cyclophilin 40 in samples. No significant cross-reactivity or interference between Mouse Cyclophilin 40 and analogues was observed. | |
10105 | |
1 Kit | |
Storage of components varies. See protocol for specific instructions. | |
Mouse |
0.19 ng/mL | |
Biological Fluids, Cell Culture Supernatant, Plasma, Serum | |
ELISA, Sandwich ELISA | |
PPIF | |
Proteases & Other Enzymes | |
Cyclophilin F, CYP3FLJ90798, Cyp-D, hCyP3, MGC117207, mitochondrial, peptidyl-prolyl cis-trans isomerase, mitochondrial, peptidylprolyl isomerase F, peptidylprolyl isomerase F (cyclophilin F) |