Learn More
Raybiotech IncSupplier Diversity Partner C-Series Mouse Cytokine Antibody Array 1000. DFS Item
Manufacturer:  Raybiotech IncSupplier Diversity Partner AAMCYT10004
A combination of Mouse Cytokine Antibody Array C3 & C4. Detects 96 Mouse Cytokines. Suitable for all liquid sample types. Semi-Quantitative, Sandwich-based, Membrane Antibody Array. Screen and compare protein expression levels with high specificity and sensitivity (pg/ml levels). No dedicated equipment required. Compatible with any chemiluminescent imaging system. View protein list on RayBiotech’s website – AAM-CYT-1000-4
Catalog No. 50-194-8694