Learn More
Raybiotech IncSupplier Diversity Partner G-Series Mouse Cytokine Antibody Array 2000. A combination of Mouse Cytokine Antibody Array G3, G4, & G5 Kit. DFS Item
Manufacturer:  Raybiotech IncSupplier Diversity Partner AAMCYTG20004
Detects 144 Mouse Cytokines. Suitable for all liquid sample types. Semi-Quantitative, Sandwich-based, Glass Slide Antibody Array Screen and compare protein expression levels with high specificity and sensitivity (pg/ml levels). The glass slide array allows for high throughput and low sample consumption; as little as 10 µl of original sample required per array. View protein list on RayBiotech’s website – AAM-CYT-G2000-4
Catalog No. 50-194-8722