Learn More
R&D Systems™ Mouse Dkk-2 Recombinant Protein Available on GSA/VA Contract for Federal Government customers only.
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.
$376.00
Specifications
Product Type Recombinant protein
Regulatory Status RUO
Recombinant Yes
Biological Activity >180pmol/min/μg
Quantity 10μg
View More Specs
Products 2
Catalog Number Mfr. No. Formulation Price Quantity & Availability  
Catalog Number Mfr. No. Formulation Price Quantity & Availability  
24-35D-KB010 Available on GSA/VA Contract for Federal Government customers only. View Documents R&D Systems™
2435DKB010
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein Each for $376.00
Add to cart
24-35D-KB010C View Documents R&D Systems™
2435DKB010/CF
Lyophilized from a 0.2μm filtered solution in PBS Each for $376.00
Add to cart
Description
Description
The Recombinant Mouse Dkk-2 Protein is derived from Sf 21 (baculovirus). The Recombinant Mouse Dkk-2 Protein has been validated for the following applications: Bioactivity.
Order Info
Ships at ambient temperature.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
Recombinant protein
Yes
10μg
>95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
<1.0 EU per 1μg of the protein by the LAL method
Spodoptera frugiperda, Sf 21 (baculovirus)-derived Ser26-Ile259, with Leu35Pro substitution and a C-terminal 10-His tag
RUO
>180pmol/min/μg
Unlabeled
Human
27 kDa
Q9QYZ8.1
Bioactivity