Learn More
Novus Biologicals™ Mouse Dopamine D1R/DRD1 ELISA Kit (Colorimetric)
ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture.
Manufacturer:  Novus Biologicals™ NBP267262
Catalog No. NBP267262
Add to cart
Description
Description
The Mouse Dopamine D1R/DRD1 ELISA Kit (Colorimetric) measures mouse Dopamine D1R/DRD1 in serum, plasma and other biological fluids.
Specifications
Specifications
0.39 - 25 ng/mL
This kit recognizes Mouse D1R in samples. No significant cross-reactivity or interference between Mouse D1R and analogues was observed.
1812
Mouse
1 Kit
Storage is content dependent.
0.23 ng/mL
ELISA
DRD1
ELISA Kit (Colorimetric)
Biological Fluids,Plasma,Serum
Dopamine D1 R/DRD1