Learn More
Novus Biologicals™ Mouse Dopamine D3R/DRD3 ELISA Kit (Colorimetric)
SDP
ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture.
Supplier:  Novus Biologicals™ NBP267939
Catalog No. NBP267939
Add to Cart
Description
Description
The Mouse Dopamine D3R/DRD3 ELISA Kit (Colorimetric) measures mouse Dopamine D3R/DRD3 in serum, plasma and other biological fluids.
Specifications
Specifications
0.16 - 10 ng/mL
This kit recognizes Mouse DRD3 in samples. No significant cross-reactivity or interference between Mouse DRD3 and analogues was observed.
Biological Fluids, Plasma, Serum
1814
Mouse
GPCR
Dopamine D3 R/DRD3
0.10 ng/mL
ELISA Kit (Colorimetric)
ELISA
DRD3
1 Kit
Storage of components varies. See protocol for specific instructions.
Recommendations
Recommendations