Learn More
Novus Biologicals™ Mouse Dopamine Receptor D4 ELISA Kit (Colorimetric)
SDP
ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture.
Supplier:  Novus Biologicals™ NBP267942
Catalog No. NBP267942
Add to Cart
Description
Description
The Mouse Dopamine Receptor D4 ELISA Kit (Colorimetric) measures mouse Dopamine Receptor D4 in serum, plasma and other biological fluids.
Specifications
Specifications
0.16 - 10 ng/mL
This kit recognizes Mouse DRD4 in samples. No significant cross-reactivity or interference between Mouse DRD4 and analogues was observed.
Biological Fluids, Plasma, Serum
1815
Mouse
GPCR
Dopamine Receptor D4
0.10 ng/mL
ELISA Kit (Colorimetric)
ELISA
drd4
1 Kit
Storage of components varies. See protocol for specific instructions.
Product Suggestions
Product Suggestions