Learn More
Abnova™ Mouse Ebi3 (Q6EAL8) Recombinant Protein
Used for Func, SDS-PAGE
Manufacturer:  Abnova™ P4622
Catalog No. 89-942-706
Add to cart
Description
Description
Sequence: MALVALSQPRVQCHASRYPVAVDCSWTPLQAPNSTRSTSFIATYRLGVATQQQSQPCLQRSPQASRCTIPDVHLFSTVPYMLNVTAVHPGGASSSLLAFVAERIIKPDPPEGVRLRTAGQRLQVLWHPPASWPFPDIFSLKYRLRYRRRGASHFRQVGPIEATTFTLRNSKPHAKYCIQVSAQDLTDYGKPSDWSLPGQVESAPHKP
Specifications
Specifications
Q6EAL8
Lyophilized
50498
Ebi3 (Mouse) Recombinant Protein
1 ug/lane in 4-20% Tris-Glycine gel Stained with Coomassie Blue
MALVALSQPRVQCHASRYPVAVDCSWTPLQAPNSTRSTSFIATYRLGVATQQQSQPCLQRSPQASRCTIPDVHLFSTVPYMLNVTAVHPGGASSSLLAFVAERIIKPDPPEGVRLRTAGQRLQVLWHPPASWPFPDIFSLKYRLRYRRRGASHFRQVGPIEATTFTLRNSKPHAKYCIQVSAQDLTDYGKPSDWSLPGQVESAPHKP
RUO
EBI-3|IL-27
Ebi3
Escherichia coli
None
Functional Study, Sodium Dodecyl Sulfate–Polyacrylamide Gel Electrophoresis
Lyophilized
22.9kDa
Escherichia coli expression system
10 ug
Store at -20°C on dry atmosphere. After reconstitution with sterilized water, store at -20°C. Aliquot to avoid repeated freezing and thawing.
<0.1 EU/μg
Ebi3
Escherichia coli expression system
Yes