Learn More
Pinnacle Technology IncSupplier Diversity Partner MOUSE EEG/EMG HEADMOUNT DFS Item
Manufacturer:  Pinnacle Technology IncSupplier Diversity Partner 8201
8mm x 5mm; weight 0.22g; EMG leads 11 mm. Connects via a friction fitting to a Pinnacle Technology preamplifier.
Catalog No. NC0117711