Learn More
Raybiotech IncSupplier Diversity Partner Mouse EGFR/ErbB1 ELISA Kit DFS Item
Manufacturer:  Raybiotech IncSupplier Diversity Partner ELMEGFR1
For Serum, Plasma, and Cell Culture Supernatants. Sensitivity: 0.2 ng/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: This ELISA antibody pair recognizes Mouse EGFR.
Catalog No. 50-194-9972