Learn More
Novus Biologicals™ Mouse ENPP-2/Autotaxin ELISA Kit (Colorimetric)
Supplier: Novus Biologicals™ NBP326057
Specifications
72 to 2500 ng/mL | |
ELISA Kit (Colorimetric) | |
This assay has high sensitivity and excellent specificity for detection of mouse ENPP-2/Autotaxin. No significant cross-reactivity or interference between mouse ENPP-2/Autotaxin and analogues was observed. | |
5168 | |
1 Kit | |
Storage of components varies. See protocol for specific instructions. | |
Mouse |
36 ng/mL | |
Serum, Plasma, Tissue Homogenates | |
ELISA, Sandwich ELISA | |
ENPP2 | |
Cardiovascular Biology | |
ATXFLJ26803, ATX-X, AUTOTAXIN, autotaxin-t, EC 3.1.4.39, ectonucleotide pyrophosphatase/phosphodiesterase 2, ectonucleotide pyrophosphatase/phosphodiesterase family member 2, E-NPP 2, Extracellular lysophospholipase D, LysoPLD, PD-IALPHA, PDNP2NPP2, phosphodiesterase I/nucleotide pyrophosphatase 2, plasma lysophospholipase D |