Learn More
Raybiotech IncSupplier Diversity Partner Mouse FABP4/A-FABP ELISA Kit DFS Item
Manufacturer:  Raybiotech IncSupplier Diversity Partner ELMFABP41
For Serum, Plasma, and Cell Culture Supernatants. Sensitivity: 14 pg/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: This ELISA antibody pair recognizes Mouse FABP4.
Catalog No. 50-194-9983