Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Gibco™ Mouse FGF-10 Recombinant Protein, PeproTech®
E. coli Recombinant Protein
$92.25 - $682.00
Specifications
Accession Number | O35565 |
---|---|
For Use With (Application) | Functional Assay |
Formulation | protein with no preservative |
Gene ID (Entrez) | 14165 |
Molecular Weight (g/mol) | 17 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
45061100UG
|
Gibco™
45061100UG |
100 μg |
Each for $682.00
|
|
|||||
450-61-25UG
|
Gibco™
4506125UG |
25 μg |
Each for $223.00
|
|
|||||
450-61-5UG
|
Gibco™
450615UG |
5 μg |
Each for $92.25
|
|
|||||
Description
FGF10 is a member of the fibroblast growth factor (FGF) family. FGF family members possess broad mitogenic and cell survival activities, and are involved in a variety of biological processes, including embryonic development, cell growth, morphogenesis, tissue repair, tumor growth and invasion. This protein exhibits mitogenic activity for keratinizing epidermal cells, but essentially no activity for fibroblasts, which is similar to the biological activity of FGF7. Studies of the mouse homolog of suggested that this gene is required for embryonic epidermal morphogenesis including brain development, lung morphogenesis, and initiation of lim bud formation. This protein is also implicated to be a primary factor in the process of wound healing.Specifications
O35565 | |
protein with no preservative | |
17 | |
E. coli | |
RUO | |
AEY17; BB213776; FGF; Fgf10; FGF-10; Fibroblast growth factor; Fibroblast growth factor 10; Gsfaey17; Keratinocyte growth factor 2; produced by fibroblasts of urinary bladder lamina propria | |
Mouse | |
Recombinant | |
Lyophilized |
Functional Assay | |
14165 | |
Mouse FGF-10 | |
-20°C | |
<1 EU/μg | |
FGF10 | |
Unconjugated | |
SSAGRHVRSY NHLQGDVRWR RLFSFTKYFL TIEKNGKVSG TKNEDCPYSV LEITSVEIGV VAVKAINSNY YLAMNKKGKL YGSKEFNNDC KLKERIEENG YNTYASFNWQ HNGRQMYVAL NGKGAPRRGQ KTRRKNTSAH FLPMTIQT | |
≥ 95% by SDS-PAGE gel and HPLC analyses. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title