Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Abnova™ Mouse Fgf1 (NP_034327, 16 a.a. - 155 a.a.) Partial Recombinant Protein with His-tag
Used for Func, SDS-PAGE
Supplier: Abnova™ P4536
Description
Sequence: MGSSHHHHHHSSGLVPRGSHMFNLPLGNYKKPKLLYCSNGGHFLRILPDGTVDGTRDRSDQHIQLQLSAESAGEVYIKGTETGQYLAMDTEGLLYGSQTPNEECLFLERLEENHYNTYTSKKHAEKNWFVGLKKNGSCKRGPRTHYGQKAILFLPLPVSSDSpecifications
NP_034327 | |
Functional Study, SDS-PAGE | |
14164 | |
Fgf1 (Mouse) Recombinant Protein | |
15% SDS-PAGE Stained with Coomassie Blue | |
MGSSHHHHHHSSGLVPRGSHMFNLPLGNYKKPKLLYCSNGGHFLRILPDGTVDGTRDRSDQHIQLQLSAESAGEVYIKGTETGQYLAMDTEGLLYGSQTPNEECLFLERLEENHYNTYTSKKHAEKNWFVGLKKNGSCKRGPRTHYGQKAILFLPLPVSSD | |
RUO | |
Fgf1 | |
E. coli | |
His | |
Liquid |
1mg/mL | |
Liquid | |
18kDa | |
Escherichia coli expression system | |
100 ug | |
Store at -20°C. For long term storage store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
Dffrx/Fam/Fgf-1/Fgfa | |
Fgf1 | |
Recombinant | |
Escherichia coli expression system | |
>90% by SDS - PAGE |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction