Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Abnova™ Mouse Fgf9 (P54130, 205 amino acids) Partial Recombinant Protein
Used for Func, SDS-PAGE
Supplier: Abnova™ P4448
Specifications
P54130 | |
Lyophilized | |
23.3kDa | |
Escherichia coli expression system | |
Store at -20°C on dry atmosphere. After reconstitution with sterilized water, store at -20°C. Aliquot to avoid repeated freezing and thawing. | |
<0.1ng/μg | |
Fgf9 | |
E. coli | |
None | |
Lyophilized |
Functional Study, SDS-PAGE | |
14180 | |
Fgf9 (Mouse) Recombinant Protein | |
10 ug | |
RUO | |
Fgf9 | |
The ED50 determined by the dose-dependent stimulation of thymidine uptake by BaF3 cells expressing FGF receptors is < 0.5ng/mL, corresponding to a specific activity of > 2 x 106 units/mg. | |
Recombinant | |
Escherichia coli expression system | |
≥98% by SDS-PAGE and HPLC |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction