Learn More
Abnova™ Mouse Fgf9 (P54130) Recombinant Protein
Used for Func, SDS-PAGE
Manufacturer:  Abnova™ P4593
Catalog No. 89-944-502
Add to cart
Description
Description
Sequence: PLGEVGSYFGVQDAVPFGNVPVLPVDSPVLLNDHLGQSEAGGLPRGPAVTDLDHLKGILRRRQLYCRTGFHLEIFPNGTIQGTRKDHSRFGILEFISIAVGLVSIRGVDSGLYLGMNEKGELYGSEKLTQECVFREQFEENWYNTYSSNLYKHVDTGRRYYVALNKDGTPREGTRTKRHQKFTHFLPRPVDPDKVPELYKDILSQS
Specifications
Specifications
P54130
Lyophilized
14180
Fgf9 (Mouse) Recombinant Protein
1 ug/lane in 4-20% Tris-Glycine gel Stained with Coomassie Blue
MPLGEVGSYFGVQDAVPFGNVPVLPVDSPVLLNDHLGQSEAGGLPRGPAVTDLDHLKGILRRRQLYCRTGFHLEIFPNGTIQGTRKDHSRFGILEFISIAVGLVSIRGVDSGLYLGMNEKGELYGSEKLTQECVFREQFEENWYNTYSSNLYKHVDTGRRYYVALNKDGTPREGTRTKRHQKFTHFLPRPVDPDKVPELYKDILSQS
RUO
Fgf9
Escherichia coli expression system
Escherichia coli
None
Functional Study, Sodium Dodecyl Sulfate–Polyacrylamide Gel Electrophoresis
Lyophilized
23.3kDa
Escherichia coli expression system
10 ug
Store at -20°C on dry atmosphere. After reconstitution with sterilized water, store at -20°C. Aliquot to avoid repeated freezing and thawing.
<0.1 EU/μg
Fgf9
The activity is determined by the dose-dependent proliferation of murine BALB/c 3T3 cells. The expected ED50 for this effect is 0.9-1.4ng/mL.
Yes