Learn More
Gibco™ Mouse gAcrp30 Recombinant Protein, PeproTech®
SDP
E. coli Recombinant Protein
Supplier:  Gibco™ 450271MG
Encompass
 View more versions of this product
Catalog No. 450-27-1MG
Add to Cart
This item is not returnable. View return policy
Description
Description
gAcrp30 is a naturally occurring globular protein obtained by proteolytic processing of adiponectin. Adiponectin is produced and secreted exclusively by adipocytes, and is a relatively abundant plasma protein, accounting for up to 0.05% of total serum protein. Like Adiponectin, gAcrp30 is capable of decreasing hyperglycemia and reversing insulin resistance. Additionally, gAcrp30 has been shown to be an important factor in promoting fat loss by signaling muscle to absorb and burn Free-Fatty Acids (FFAs). The signaling receptors for adiponectin and gAcrp30 have recently been identified and named AdipoR1 and AdipoR2. AdipoR2 is predominantly expressed in the liver.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Q60994
protein with no preservative
16.6
2 x 500 μg
-20°C
<1 EU/μg
ADIPOQ
Unconjugated
MKGEPGEAAY MYRSAFSVGL ETRVTVPNVP IRFTKIFYNQ QNHYDGSTGK FYCNIPGLYY FSYHITVYMK DVKVSLFKKD KAVLFTYDQY QEKNVDQASG SVLLHLEVGD QVWLQVYGDG DHNGLYADNV NDSTFTGFLL YHDTN
≥ 98% by SDS-PAGE gel and HPLC analyses.
Functional Assay
11450
Mouse gAcrp30
E. coli
RUO
gAcrp30
Mouse
Recombinant
Lyophilized
Product Suggestions
Product Suggestions