Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Gibco™ Mouse gAcrp30 Recombinant Protein, PeproTech®
E. coli Recombinant Protein
$89.50 - $3325.00
Specifications
Accession Number | Q60994 |
---|---|
For Use With (Application) | Functional Assay |
Gene ID (Entrez) | 11450 |
Molecular Weight (g/mol) | 16.6 |
Name | Mouse gAcrp30 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
45027100UG
|
Gibco™
45027100UG |
100 μg |
Each for $666.00
|
|
|||||
45-027-01M
|
Gibco™
4502701M |
2 x 500 μg |
Each for $3,325.00
|
|
|||||
45027250UG
|
Gibco™
45027250UG |
250 μg |
Each for $1,164.00
|
|
|||||
450-27-25UG
|
Gibco™
4502725UG |
25 μg |
Each for $217.00
|
|
|||||
45027500UG
|
Gibco™
45027500UG |
500 μg |
Each for $1,996.00
|
|
|||||
450-27-50UG
|
Gibco™
4502750UG |
50 μg |
Each for $400.00
|
|
|||||
450-27-5UG
|
Gibco™
450275UG |
5 μg |
Each for $89.50
|
|
|||||
Description
gAcrp30 is a naturally occurring globular protein obtained by proteolytic processing of adiponectin. Adiponectin is produced and secreted exclusively by adipocytes, and is a relatively abundant plasma protein, accounting for up to 0.05% of total serum protein. Like Adiponectin, gAcrp30 is capable of decreasing hyperglycemia and reversing insulin resistance. Additionally, gAcrp30 has been shown to be an important factor in promoting fat loss by signaling muscle to absorb and burn Free-Fatty Acids (FFAs). The signaling receptors for adiponectin and gAcrp30 have recently been identified and named AdipoR1 and AdipoR2. AdipoR2 is predominantly expressed in the liver.Specifications
Q60994 | |
11450 | |
Mouse gAcrp30 | |
-20°C | |
<1 EU/μg | |
ADIPOQ | |
Unconjugated | |
MKGEPGEAAY MYRSAFSVGL ETRVTVPNVP IRFTKIFYNQ QNHYDGSTGK FYCNIPGLYY FSYHITVYMK DVKVSLFKKD KAVLFTYDQY QEKNVDQASG SVLLHLEVGD QVWLQVYGDG DHNGLYADNV NDSTFTGFLL YHDTN | |
≥ 98% by SDS-PAGE gel and HPLC analyses. |
Functional Assay | |
16.6 | |
E. coli | |
RUO | |
gAcrp30 | |
Mouse | |
Recombinant | |
Lyophilized |
Spot an opportunity for improvement?
Product Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title