Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Novus Biologicals™ Mouse GAD Autoantibody (IgG) ELISA Kit (Colorimetric)
Colorimetric ELISA Kit for study of Apolipoproteins, Lipid transport
Supplier: Novus Biologicals™ NBP2600251KIT
Description
The Mouse GAD Autoantibody (IgG) ELISA Kit (Colorimetric) measures mouse GAD Autoantibody (IgG) in serum, plasma.Specifications
Qualitative | |
Plasma, Serum | |
Mouse | |
Coated assay plate, Negative Control, Positive Control, Sample Diluents, HRP- conjugate, Wash Buffer (20X concentrate), Substrate A, Substrate B, Stop Solution, Adhesive Strip (For 96 wells), Instruction manual | |
RUO | |
50 to 100 μL | |
GAD Autoantibody, Glutamic acid decarboxylase autoantibody |
ELISA Kit (Colorimetric) | |
Detection of mouse glutamic acid decarboxylase (GAD) autoantibody (IgG) | |
ELISA | |
1 Kit | |
Immune Dysfunction | |
Store the unopened product at 2° to 8°C. Protect from light. Do not use past expiration date. | |
GAD Autoantibody (IgG) |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction